1-(3-Amino-4-morpholinophenyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

045459-500MG 045459-1G 045459-5G
101870-932EA 556.92 USD
101870-932 101870-934 101870-936
1-(3-Amino-4-morpholinophenyl)-1-ethanone ≥95%
1-(3-Amino-4-morpholinophenyl)-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD00116066
CAS Number: 217489-78-6

Order Now

Specification Test Results

Melting point 162-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR