Methyl-4-formyl-1-methyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate

045457-500MG 045457-1G
101870-922EA 392.31 USD
101870-922 101870-924
Methyl-4-formyl-1-methyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-formyl-1-methyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.17 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD01571196
CAS Number: 67858-47-3

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR