2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045455-500MG 045455-1G 045455-5G
101870-910EA 556.92 USD
101870-910 101870-912 101870-914
2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylic acid ≥95%
2-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₈ClNO₃S
Melting Pt: 130…131 °C
MDL Number: MFCD05663801
CAS Number: 886360-91-4

Order Now

Specification Test Results

Melting point 130-131°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR