1-{[4-(Acetylamino)phenyl]sulfonyl}-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

045454-500MG 045454-1G 045454-5G
101870-904EA 181.54 USD
101870-904 101870-906 101870-908
1-{[4-(Acetylamino)phenyl]sulfonyl}-4-piperidinecarboxylic acid ≥95%
1-{[4-(Acetylamino)phenyl]sulfonyl}-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₅S
MW: 326.37 g/mol
Melting Pt: 215…217 °C
MDL Number: MFCD02081043
CAS Number: 314744-44-0

Order Now

Specification Test Results

Melting point 215-217°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR