4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4,7,8-Tetrahydroquinoline-2,5(1H,6H)-dione

045452-500MG 045452-1G 045452-5G
101870-892EA 278.46 USD
101870-892 101870-894 101870-896
4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione ≥95%
4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 194…195 °C
MDL Number: MFCD01111799
CAS Number: 5057-12-5

Order Now

Specification Test Results

Melting point 196-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR