Methyl-1,3-benzodioxole-5-carboxylate ≥95%

Supplier: Matrix Scientific

045449-500MG 045449-1G 045449-5G
101870-882EA 181.54 USD
101870-882 101870-884 101870-886
Methyl-1,3-benzodioxole-5-carboxylate ≥95%
Methyl-1,3-benzodioxole-5-carboxylate

Hazard: Irritant

Formula: C₉H₈O₄
Melting Pt: 50 °C
MDL Number: MFCD00030273
CAS Number: 326-56-7

Order Now

Specification Test Results

Melting point 50°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR