tert-Butyl-4-[4-(methoxycarbonyl)-2-oxo-1-pyrrolidinyl]tetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
4-[4-(Methoxycarbonyl)-2-oxopyrrolidin-1-yl]piperidine, N-BOC protected
045445-500MG
045445-1G
045445-5G
101870-860EA
375.38
USD
101870-860
101870-862
101870-864
tert-Butyl-4-[4-(methoxycarbonyl)-2-oxo-1-pyrrolidinyl]tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-[4-(methoxycarbonyl)-2-oxo-1-pyrrolidinyl]tetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₆N₂O₅ MW: 326.4 g/mol Melting Pt: 98…100 °C |
MDL Number:
MFCD08689666 CAS Number: 937601-48-4 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...