Methyl-5-chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-chloro-2-oxo-1,2-dihydropyridine-3-carboxylate

045443-500MG 045443-1G 045443-5G
101870-850EA 375.38 USD
101870-850 101870-852 101870-854
Methyl-5-chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylate ≥95%
Methyl-5-chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 171…174 °C
MDL Number: MFCD04113615
CAS Number: 886360-77-6

Order Now

Specification Test Results

Melting point 171-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR