Ethyl-4-oxo-2-(2-toluidino)-4,5-dihydro-3-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045440-500MG 045440-1G 045440-5G
101870-838EA 375.38 USD
101870-838 101870-840 101870-842
Ethyl-4-oxo-2-(2-toluidino)-4,5-dihydro-3-thiophenecarboxylate ≥95%
Ethyl-4-oxo-2-(2-toluidino)-4,5-dihydro-3-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₅NO₃S
Melting Pt: 139…141 °C
MDL Number: MFCD03791239
CAS Number: 886360-76-5

Order Now

Specification Test Results

Melting point 139-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR