Methyl-2-(3-formylphenoxy)-5-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

045439-500MG 045439-1G 045439-5G
101870-832EA 278.46 USD
101870-832 101870-834 101870-836
Methyl-2-(3-formylphenoxy)-5-nitrobenzenecarboxylate ≥95%
Methyl-2-(3-formylphenoxy)-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₁NO₆
Melting Pt: 103…105 °C
MDL Number: MFCD03844856
CAS Number: 886360-75-4

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR