Methyl-5-bromo-6-hydroxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-6-hydroxynicotinate

045438-500MG 045438-1G 045438-5G
101870-826EA 375.38 USD
101870-826 101870-828 101870-830
Methyl-5-bromo-6-hydroxynicotinate ≥95%
Methyl-5-bromo-6-hydroxynicotinate

Hazard: Irritant

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Melting Pt: 225-227°C
MDL Number: MFCD08235247
CAS Number: 381247-99-0

Order Now

Specification Test Results

Melting point 225-227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR