Methyl-2-(4-benzylpiperazino)-5-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

045434-500MG 045434-1G 045434-5G
101870-812EA 375.38 USD
101870-812 101870-814 101870-816
Methyl-2-(4-benzylpiperazino)-5-nitrobenzenecarboxylate ≥95%
Methyl-2-(4-benzylpiperazino)-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₉H₂₁N₃O₄
Melting Pt: 113…115 °C
MDL Number: MFCD02643034
CAS Number: 381195-04-6

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR