Ethyl-1-[2-(methoxycarbonyl)-4-nitrophenyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

045433-500MG 045433-1G 045433-5G
101870-806EA 375.38 USD
101870-806 101870-808 101870-810
Ethyl-1-[2-(methoxycarbonyl)-4-nitrophenyl]-4-piperidinecarboxylate ≥95%
Ethyl-1-[2-(methoxycarbonyl)-4-nitrophenyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₆
Melting Pt: 117…119 °C
MDL Number: MFCD05021845
CAS Number: 886360-74-3

Order Now

Specification Test Results

Melting point 117-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR