Ethyl-2-anilino-4-chloro-5-formyl-3-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045432-500MG 045432-1G 045432-5G
101870-800EA 278.46 USD
101870-800 101870-802 101870-804
Ethyl-2-anilino-4-chloro-5-formyl-3-thiophenecarboxylate ≥95%
Ethyl-2-anilino-4-chloro-5-formyl-3-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₂ClNO₃S
Melting Pt: 118…120 °C
MDL Number: MFCD02181910
CAS Number: 78267-24-0

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR