3,6-Dimethoxy-2-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

045429-500MG 045429-1G 045429-5G
101870-790EA 181.54 USD
101870-790 101870-792 101870-794
3,6-Dimethoxy-2-nitrobenzaldehyde ≥95%
3,6-Dimethoxy-2-nitrobenzaldehyde

Hazard: Irritant

Formula: C₉H₉NO₅
Melting Pt: 162-164°C
MDL Number: MFCD00091893
CAS Number: 1206-55-9

Order Now

Specification Test Results

Melting point 162-164°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR