6-(1-Pyrrolidinyl)nicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(Pyrrolidin-1-yl)pyridine-3-carboxylic acid, 6-Pyrrolidin-1-yl-nicotinic acid

045428-500MG 045428-1G 045428-5G
101870-784EA 112.31 USD
101870-784 101870-786 101870-788
6-(1-Pyrrolidinyl)nicotinic acid ≥95%
6-(1-Pyrrolidinyl)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Melting Pt: 259…261 °C
MDL Number: MFCD03791234
CAS Number: 210963-95-4

Order Now

Specification Test Results

Melting point 259-261°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR