Ethyl-2-(2-pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

045425-500MG 045425-1G 045425-5G
101870-772EA 375.38 USD
101870-772 101870-774 101870-776
Ethyl-2-(2-pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylate ≥95%
Ethyl-2-(2-pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀F₃N₃O₂
Melting Pt: 98…100 °C
MDL Number: MFCD05668813
CAS Number: 866132-66-3

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR