1-(Benzylsulfanyl)-4-nitrobenzene ≥95%
Supplier: Matrix Scientific
045423-500MG
045423-1G
045423-5G
101870-760EA
181.54
USD
101870-760
101870-762
101870-764
1-(Benzylsulfanyl)-4-nitrobenzene ≥95%
1-(Benzylsulphanyl)-4-nitrobenzene
Hazard: Irritant
Formula:
C₁₃H₁₁NO₂S Melting Pt: 91…93 °C |
MDL Number:
MFCD00034549 CAS Number: 27691-43-6 |
Specification Test Results
Melting point | 91-93°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...