1-(Benzylsulfanyl)-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

045421-500MG 045421-1G 045421-5G
101870-748EA 181.54 USD
101870-748 101870-750 101870-752
1-(Benzylsulfanyl)-2-nitrobenzene ≥95%
1-(Benzylsulphanyl)-2-nitrobenzene

Hazard: Irritant

Formula: C₁₃H₁₁NO₂S
Melting Pt: 80…81 °C
MDL Number: MFCD00031120
CAS Number: 22057-44-9

Order Now

Specification Test Results

Melting point 80-81°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR