2-[(Carboxymethyl)amino]propanoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

045419-500MG 045419-1G 045419-5G
101870-736EA 181.54 USD
101870-736 101870-738 101870-740
2-[(Carboxymethyl)amino]propanoic acid hydrochloride ≥95%
2-[(Carboxymethyl)amino]propanoic acid hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClNO₄
Melting Pt: 238…240 °C
MDL Number: MFCD03844854
CAS Number: 33012-75-8

Order Now

Specification Test Results

Melting point 238-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR