4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid

045418-500MG 045418-1G 045418-5G
101870-730EA 78.46 USD
101870-730 101870-732 101870-734
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid ≥95%
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
Melting Pt: 196…198 °C
MDL Number: MFCD00453921
CAS Number: 15898-26-7

Order Now

Specification Test Results

Melting point 181-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR