Methyl-2-(4,5-dichloro-2-nitrophenyl)acetate ≥95%

Supplier: Matrix Scientific

045417-500MG 045417-1G 045417-5G
101870-724EA 278.46 USD
101870-724 101870-726 101870-728
Methyl-2-(4,5-dichloro-2-nitrophenyl)acetate ≥95%
Methyl-2-(4,5-dichloro-2-nitrophenyl)acetate

Hazard: Irritant

Formula: C₉H₇Cl₂NO₄
Melting Pt: 66…69 °C
MDL Number: MFCD03425813
CAS Number: 286949-63-1

Order Now

Specification Test Results

Melting point 66-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR