Ethyl-6-methyl-4-(4-methylphenyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

045416-500MG 045416-1G 045416-5G
101870-718EA 181.54 USD
101870-718 101870-720 101870-722
Ethyl-6-methyl-4-(4-methylphenyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-6-methyl-4-(4-methylphenyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₃
Melting Pt: 218…219 °C
MDL Number: MFCD00719779
CAS Number: 299949-24-9

Order Now

Specification Test Results

Melting point 218-219°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR