7-Cyclopropylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
Supplier: Matrix Scientific
045415-500MG
045415-1G
101870-714EA
278.46
USD
101870-714
101870-716
7-Cyclopropylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
7-Cyclopropylpyrazolo[1,5-a]pyrimidine-3-carbonitrile
Hazard: Irritant
Formula:
C₁₀H₈N₄ Melting Pt: 165…167 °C |
MDL Number:
MFCD03305760 CAS Number: 866132-04-9 |
Specification Test Results
Melting point | 165-167°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...