1-[(2-Aminoethyl)amino]-6-methyl-9H-xanthen-9-one ≥95%

Supplier: Matrix Scientific

045413-500MG 045413-1G 045413-5G
101870-702EA 375.38 USD
101870-702 101870-704 101870-706
1-[(2-Aminoethyl)amino]-6-methyl-9H-xanthen-9-one ≥95%
1-[(2-Aminoethyl)amino]-6-methyl-9H-xanthen-9-one

Hazard: Irritant

Formula: C₁₆H₁₆N₂O₂
Melting Pt: 178…180 °C
MDL Number: MFCD03425811
CAS Number: 359028-07-2

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR