2-Chloro-4-fluoro-5-nitroanisole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-5-fluoro-2-methoxy-4-nitrobenzene

045411-1G 045411-5G
101870-698EA 44.62 USD
101870-698 101870-700
2-Chloro-4-fluoro-5-nitroanisole ≥95%
2-Chloro-4-fluoro-5-nitroanisole

Hazard: Irritant

Formula: C₇H₅ClFNO₃
MW: 205.5700 g/mol
Melting Pt: 72-75°C
MDL Number: MFCD03425810
CAS Number: 84478-76-2

Order Now

Specification Test Results

Melting point 72-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR