1-Chloro-5-fluoro-2-[(4-fluorobenzyl)oxy]-4-nitrobenzene ≥95%

Supplier: Matrix Scientific

045408-500MG 045408-1G 045408-5G
101870-680EA 556.92 USD
101870-680 101870-682 101870-684
1-Chloro-5-fluoro-2-[(4-fluorobenzyl)oxy]-4-nitrobenzene ≥95%
1-Chloro-5-fluoro-2-[(4-fluorobenzyl)oxy]-4-nitrobenzene

Hazard: Irritant

Formula: C₁₃H₈ClF₂NO₃
Melting Pt: 163…166 °C
MDL Number: MFCD03425808
CAS Number: 439095-66-6

Order Now

Specification Test Results

Melting point 163-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR