2-(4,5-Dichloro-2-nitrophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

045407-500MG 045407-1G 045407-5G
101870-674EA 181.54 USD
101870-674 101870-676 101870-678
2-(4,5-Dichloro-2-nitrophenyl)acetic acid ≥95%
2-(4,5-Dichloro-2-nitrophenyl)acetic acid

Hazard: Irritant

Formula: C₈H₅Cl₂NO₄
Melting Pt: 127…130 °C
MDL Number: MFCD07366549
CAS Number: 37777-90-5

Order Now

Specification Test Results

Melting point 127-130°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR