1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

045405-500MG 045405-1G 045405-5G
101870-662EA 150.77 USD
101870-662 101870-664 101870-666
1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
Melting Pt: 202…204 °C
MDL Number: MFCD00829059
CAS Number: 64038-03-5

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR