3-Amino-3-cyclohexylpropanoic acid ≥95%

Supplier: Matrix Scientific

045404-500MG 045404-1G 045404-5G
101870-656EA 192.31 USD
101870-656 101870-658 101870-660
3-Amino-3-cyclohexylpropanoic acid ≥95%
3-Amino-3-cyclohexylpropanoic acid

Hazard: Irritant

Formula: C₉H₁₇NO₂
Melting Pt: 221…223 °C
MDL Number: MFCD01863250
CAS Number: 129042-71-3

Order Now

Specification Test Results

Melting point 221-223°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR