2-(2-Thienylmethylene)malononitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Thiophen-2-ylmethylene)malononitrile

045403-500MG 045403-1G 045403-5G
101870-650EA 181.54 USD
101870-650 101870-652 101870-654
2-(2-Thienylmethylene)malononitrile ≥95%
2-(2-Thienylmethylene)malononitrile

Hazard: Irritant

Formula: C₈H₄N₂S
MW: 160.2 g/mol
Melting Pt: 92…93 °C
MDL Number: MFCD00114006
CAS Number: 28162-32-5

Order Now

Specification Test Results

Melting point 92-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR