3-Methyl-1,3-thiazolane-2,4-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methylthiazolidine-2,4-dione

045402-500MG 045402-1G 045402-5G
101870-644EA 278.46 USD
101870-644 101870-646 101870-648
3-Methyl-1,3-thiazolane-2,4-dione ≥95%
3-Methyl-1,3-thiazolane-2,4-dione

Hazard: Irritant

Formula: C₄H₅NO₂S
MW: 131.15 g/mol
Melting Pt: 40…43 °C
MDL Number: MFCD02194457
CAS Number: 16312-21-3

Order Now

Specification Test Results

Melting point 40-43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR