Dimethyl-2-(4-nitrophenyl)malonate ≥95%

Supplier: Matrix Scientific

Synonyms: Dimethyl 2-(4-nitrophenyl)malonate

045399-500MG 045399-1G 045399-5G
101870-632EA 181.54 USD
101870-632 101870-634 101870-636
Dimethyl-2-(4-nitrophenyl)malonate ≥95%
Dimethyl-2-(4-nitrophenyl)malonate

Hazard: Irritant

Formula: C₁₁H₁₁NO₆
MW: 253.21 g/mol
Melting Pt: 125…127 °C
MDL Number: MFCD03425806
CAS Number: 4033-88-9

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR