Methyl-1-(4-chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

045397-500MG 045397-1G 045397-5G
101870-626EA 278.46 USD
101870-626 101870-628 101870-630
Methyl-1-(4-chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-1-(4-chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₂H₉ClN₂O₃
MW: 264.67 g/mol
Melting Pt: 186…189 °C
MDL Number: MFCD03001289
CAS Number: 453557-76-1

Order Now

Specification Test Results

Melting point 186-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR