3-[(4-Methylbenzoyl)amino]-2-thiophenecarboxylic acid ≥95%
Supplier: Matrix Scientific
045396-500MG
045396-1G
045396-5G
101870-620EA
181.54
USD
101870-620
101870-622
101870-624
3-[(4-Methylbenzoyl)amino]-2-thiophenecarboxylic acid ≥95%
3-[(4-Methylbenzoyl)amino]-2-thiophenecarboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₁NO₃S Melting Pt: 197…200 °C |
MDL Number:
MFCD02572044 CAS Number: 478080-03-4 |
Specification Test Results
Melting point | 197-200°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...