2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)-1-ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-ol

045394-500MG 045394-1G 045394-5G
101870-608EA 407.69 USD
101870-608 101870-610 101870-612
2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)-1-ethanol ≥95%
2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₀F₃NO
MW: 229.20 g/mol
Melting Pt: 72…73 °C
MDL Number: MFCD02571238
CAS Number: 116757-91-6

Order Now

Specification Test Results

Melting point 72-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR