N-(3-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

045391-500MG 045391-1G 045391-5G
101870-592EA 181.54 USD
101870-592 101870-594 101870-596
N-(3-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide ≥95%
N-(3-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₂
Melting Pt: 132…134 °C
MDL Number: MFCD02187344
CAS Number: 453557-74-9

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR