3-(Boc-amino)-3-(3-chlorophenyl)propionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-tert-Butoxycarbonylamino-3-(3-chlorophenyl)propionic acid, 3-[(tert-Butoxycarbonyl)amino]-3-(3-chlorophenyl)-propanoic acid, 3-tert-butoxycarbonylamino-3-(3-chlorophenyl)propionic acid

045387-500MG 045387-1G
101870-588EA 556.92 USD
101870-588 101870-590
3-(Boc-amino)-3-(3-chlorophenyl)propionic acid ≥95%
3-(Boc-amino)-3-(3-chlorophenyl)propionic acid

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MW: 299.76 g/mol
Melting Pt: 95…96 °C
MDL Number: MFCD02090710
CAS Number: 284493-67-0

Order Now

Specification Test Results

Melting point 95-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR