3-[(tert-Butoxycarbonyl)amino]-3-(4-isopropylphenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045386-500MG 045386-1G
101870-584EA 556.92 USD
101870-584 101870-586
3-[(tert-Butoxycarbonyl)amino]-3-(4-isopropylphenyl)propanoic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]-3-(4-isopropylphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₇H₂₅NO₄
MW: 307.39 g/mol
Melting Pt: 125…128 °C
MDL Number: MFCD03001287
CAS Number: 453557-73-8

Order Now

Specification Test Results

Melting point 125-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR