3-[(tert-Butoxycarbonyl)amino]-3-(3,4-dichlorophenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045385-500MG 045385-1G 045385-5G
101870-578EA 556.92 USD
101870-578 101870-580 101870-582
3-[(tert-Butoxycarbonyl)amino]-3-(3,4-dichlorophenyl)propanoic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]-3-(3,4-dichlorophenyl)propanoic acid

Hazard: Irritant

Formula: C₁₄H₁₇Cl₂NO₄
Melting Pt: 143…145 °C
MDL Number: MFCD02090707
CAS Number: 193633-52-2

Order Now

Specification Test Results

Melting point 143-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR