3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(3,4-dimethoxyphenyl)propanoic acid

045381-500MG 045381-1G
101870-566EA 461.54 USD
101870-566 101870-568
3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionic acid ≥95%
3-(Boc-amino)-3-(3,4-dimethoxyphenyl)propionic acid

Hazard: Irritant

Formula: C₁₆H₂₃NO₆
MW: 325.35 g/mol
Melting Pt: 130…131 °C
MDL Number: MFCD02090700
CAS Number: 284492-37-1

Order Now

Specification Test Results

Melting point 130-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR