3-[(tert-Butoxycarbonyl)amino]-3-(4-ethoxyphenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045380-500MG 045380-1G
101870-562EA 556.92 USD
101870-562 101870-564
3-[(tert-Butoxycarbonyl)amino]-3-(4-ethoxyphenyl)propanoic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]-3-(4-ethoxyphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₆H₂₃NO₅
Melting Pt: 164…166 °C
MDL Number: MFCD02278401
CAS Number: 453557-72-7

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR