3-(4-Bromophenyl)-3-[(tert-butoxycarbonyl)amino]propanoic acid ≥95%

Supplier: Matrix Scientific

045376-500MG 045376-1G
101870-552EA 269.23 USD
101870-552 101870-554
3-(4-Bromophenyl)-3-[(tert-butoxycarbonyl)amino]propanoic acid ≥95%
3-(4-Bromophenyl)-3-[(tert-butoxycarbonyl)amino]propanoic acid

Hazard: Irritant

Formula: C₁₄H₁₈BrNO₄
Melting Pt: 195…196 °C
MDL Number: MFCD02093998
CAS Number: 282524-86-1

Order Now

Specification Test Results

Melting point 195-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR