3-Amino-3-(3,4-dichlorophenyl)propionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-3-(3,4-dichlorophenyl)propanoic acid

045374-500MG 045374-1G 045374-5G
101870-542EA 269.23 USD
101870-542 101870-544 101870-546
3-Amino-3-(3,4-dichlorophenyl)propionic acid ≥95%
3-Amino-3-(3,4-dichlorophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
MW: 234.0793 g/mol
Melting Pt: 224-226°C
MDL Number: MFCD00462884
CAS Number: 117391-57-8

Order Now

Specification Test Results

Melting point 224-226°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR