3-Amino-3-(2-bromophenyl)propionic acid ≥95%

Supplier: Matrix Scientific

045373-500MG 045373-1G 045373-5G
101870-536EA 278.46 USD
101870-536 101870-538 101870-540
3-Amino-3-(2-bromophenyl)propionic acid ≥95%
3-Amino-3-(2-bromophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₁₀BrNO₂
Melting Pt: 221…223 °C
MDL Number: MFCD01871297
CAS Number: 117391-48-7

Order Now

Specification Test Results

Melting point 221-223°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR