Methyl-3-amino-3-(4-bromophenyl)propanoate ≥95%
Supplier: Matrix Scientific
045372-500MG
045372-1G
101870-532EA
461.54
USD
101870-532
101870-534
Methyl-3-amino-3-(4-bromophenyl)propanoate ≥95%
Methyl-3-amino-3-(4-bromophenyl)propanoate
Hazard: Irritant
Formula:
C₁₀H₁₂BrNO₂ MW: 258.12 g/mol Melting Pt: 48…49 °C |
MDL Number:
MFCD02286059 CAS Number: 453557-71-6 |
Specification Test Results
Melting point | 48-49°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...