2,2,2-Trichloro-1-[4-(3-methoxybenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

045362-500MG 045362-1G
101870-502EA 556.92 USD
101870-502 101870-504
2,2,2-Trichloro-1-[4-(3-methoxybenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%
2,2,2-Trichloro-1-[4-(3-methoxybenzoyl)-1H-pyrrol-2-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₄H₁₀Cl₃NO₃
Melting Pt: 135…137 °C
MDL Number: MFCD03001284
CAS Number: 453557-69-2

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR