2,2,2-Trichloro-1-[4-(4-methylbenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

045360-500MG 045360-1G
101870-492EA 556.92 USD
101870-492 101870-494
2,2,2-Trichloro-1-[4-(4-methylbenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%
2,2,2-Trichloro-1-[4-(4-methylbenzoyl)-1H-pyrrol-2-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₄H₁₀Cl₃NO₂
Melting Pt: 177…179 °C
MDL Number: MFCD03001282
CAS Number: 453557-68-1

Order Now

Specification Test Results

Melting point 177-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR