2,2,2-Trichloro-1-[4-(4-fluorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4-Fluorobenzoyl)-2-(trichloroacetyl)-1H-pyrrole

045358-500MG 045358-1G
101870-484EA 556.92 USD
101870-484 101870-486
2,2,2-Trichloro-1-[4-(4-fluorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone ≥95%
2,2,2-Trichloro-1-[4-(4-fluorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₃H₇Cl₃FNO₂
MW: 334.56 g/mol
Melting Pt: 202…204 °C
MDL Number: MFCD03001281
CAS Number: 453557-67-0

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR