Ethyl naphtho[2,1-b]furan-2-carboxylate ≥95%
Supplier: Matrix Scientific
045357-500MG
045357-1G
045357-5G
101870-478EA
556.92
USD
101870-478
101870-480
101870-482
Ethyl naphtho[2,1-b]furan-2-carboxylate ≥95%
Ethyl naphtho[2,1-b]furan-2-carboxylate
Hazard: Irritant
Formula:
C₁₅H₁₂O₃ Melting Pt: 113…114 °C |
MDL Number:
MFCD03001280 CAS Number: 32730-03-3 |
Specification Test Results
Melting point | 113-114°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...